Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaSep™ PREMIUM Spherical Flash Cartridges, Silica-Based, C18 Monomeric, 25 μm, 90 Å

SiliaSep™ PREMIUM Spherical Flash Cartridges, Silica-Based, C18 Monomeric, 25 μm, 90 Å (4 g to 1600 g) are packed with spherical hydrophobic C18 sorbent (17% carbon) for purification of various organic compounds.

$84.70 - $10140.00

Specifications

For Use With (Application) Purification of low to medium polarity compounds
Material Polypropylene
Type Flash Cartridge
Particle Size 25 μm
Endcapped Yes
View More Specs

Products 18
Catalog Number Mfr. No. Column Type Max. Pressure Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Column Type Max. Pressure Quantity Price Quantity & Availability  
01-183-450
View Documents
SiliCycle™
FLH03295DAISO04
4 g Flash Cartridge 225 psi/16 bar 2 Pk.
Pack of 2 for $84.70
Add to Cart
 
01-183-451
View Documents
SiliCycle™
FLH03295DAISO04B
4 g Flash Cartridge 225 psi/16 bar 10 Pk.
Pack of 10 for $301.00
Add to Cart
 
01-183-452
View Documents
SiliCycle™
FLH03295DAISO12
12 g Flash Cartridge 225 psi/16 bar 1 Pk.
Pack for $122.80
Add to Cart
 
01-183-453
View Documents
SiliCycle™
FLH03295DAISO12B
12 g Flash Cartridge 225 psi/16 bar 10 Pk.
Pack of 10 for $958.00
Add to Cart
 
01-183-454
View Documents
SiliCycle™
FLH03295DAISO25
25 g Flash Cartridge 225 psi/16 bar 1 Pk.
Pack for $244.00
Add to Cart
 
01-183-455
View Documents
SiliCycle™
FLH03295DAISO25B
25 g Flash Cartridge 225 psi/16 bar 10 Pk.
Pack of 10 for $2,060.00
Add to Cart
 
01-183-456
View Documents
SiliCycle™
FLH03295DAISO40
40 g Flash Cartridge 225 psi/16 bar 1 Pk.
Pack for $365.50
Add to Cart
 
01-183-457
View Documents
SiliCycle™
FLH03295DAISO40B
40 g Flash Cartridge 225 psi/16 bar 5 Pk.
Pack of 5 for $1,544.00
Add to Cart
 
01-183-458
View Documents
SiliCycle™
FLH03295DAISO80
80 g Flash Cartridge 225 psi/16 bar 1 Pk.
Pack for $581.50
Add to Cart
 
01-183-459
View Documents
SiliCycle™
FLH03295DAISO80B
80 g Flash Cartridge 225 psi/16 bar 5 Pk.
Pack of 5 for $2,470.00
Add to Cart
 
01-183-443
View Documents
SiliCycle™
FLH03295DAIS120
120 g Flash Cartridge 205 psi/13 bar 1 Pk.
Pack for $845.00
Add to Cart
 
01-183-444
View Documents
SiliCycle™
FLH03295DAIS120B
120 g Flash Cartridge 205 psi/13 bar 5 Pk.
Pack of 5 for $3,740.00
Add to Cart
 
01-183-445
View Documents
SiliCycle™
FLH03295DAIS220
220 g Flash Cartridge 160 psi/11 bar 1 Pk.
Pack for $1,583.00
Add to Cart
 
01-183-446
View Documents
SiliCycle™
FLH03295DAIS220B
220 g Flash Cartridge 160 psi/11 bar 4 Pk.
Pack of 4 for $5,400.00
Add to Cart
 
01-183-447
View Documents
SiliCycle™
FLH03295DAIS330
330 g Flash Cartridge 160 psi/11 bar 1 Pk.
Pack for $2,240.00
Add to Cart
 
01-183-448
View Documents
SiliCycle™
FLH03295DAIS330B
330 g Flash Cartridge 160 psi/11 bar 4 Pk.
Pack of 4 for $7,350.00
Add to Cart
 
01-183-449
View Documents
SiliCycle™
FLH03295DAIS750
800 g Flash Cartridge 125 psi/8 bar 1 Pk.
Pack for $4,830.00
Add to Cart
 
01-183-442
View Documents
SiliCycle™
FLH03295DAI1500
1600 g Flash Cartridge 100 psi/7 bar 1 Pk.
Pack for $10,140.00
Add to Cart
 
Description

Description

  • Directly compatible with a variety of flash systems (slip tip connection type, but can be used with many luer lock systems)
  • Pre-packed disposable flash cartridges
  • Unique one-piece polypropylene cartridge for high pressure and solvent resistance
  • Leak-free guaranteed
  • Made with high quality spherical silica (25 μm, 90 Å)
  • Tight particle size distribution
  • Low fines
  • Neutral pH
  • Controlled water content
  • Very diversified formats (4, 12, 25, 40, 80, 120, 220, 330, 800, and 1600 g)
  • Effective packing technology
  • Good resolution, no tailing
  • Stringent quality control (ISO 9001:2015 Registered)
  • Lot-to-lot reproducibility
  • For 800 g and 1600 g XL formats, an adapter may be needed to connect the cartridge to the system
Specifications

Specifications

Purification of low to medium polarity compounds
Flash Cartridge
Yes
Spherical
90 Å
C18 (Octadecyl, ODS)
Polypropylene
25 μm
PREMIUM (Spherical Silica)
2 to 10
SiliaSep™ PREMIUM
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit