Learn More
SiliCycle™ SiliaPrepX™ SPE Cartridges - WCX
SPE Cartridges packed with a polymeric weak cation exchange (WCX) phase, for extraction of strong basic compounds.
$208.50 - $2650.00
Specifications
Adsorbent Type | Polymer |
---|---|
Carbon Load | 0.85 |
For Use With (Application) | Extraction of strongly basic compounds, highly stable in organic solvents |
For Use With (Equipment) | Vacuum Manifolds or Automated SPE Equipment |
Particle Size | 85 μm |
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
01-184-805
|
SiliCycle™
SPEP001501AA |
30 mg | 1 mL | 100 Pk. |
Pack of 100 for $295.00
|
|
|||
01-184-588
|
SiliCycle™
SPEP001501AAL |
30 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,394.00
|
|
|||
01-184-857
|
SiliCycle™
SPEP001501AAM |
30 mg | 1 mL | 1000 Pk. |
Pack of 1000 for $2,650.00
|
|
|||
01-184-592
|
SiliCycle™
SPEP001503AA |
30 mg | 3 mL | 50 Pk. |
Pack of 50 for $208.50
|
|
|||
01-184-593
|
SiliCycle™
SPEP001503AAJ |
30 mg | 3 mL | 200 Pk. |
Pack of 200 for $744.00
|
|
|||
01-184-589
|
SiliCycle™
SPEP001501BB |
60 mg | 1 mL | 100 Pk. |
Pack of 100 for $316.50
|
|
|||
01-184-590
|
SiliCycle™
SPEP001501BBL |
60 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,498.00
|
|
|||
01-184-594
|
SiliCycle™
SPEP001503BB |
60 mg | 3 mL | 50 Pk. |
Pack of 50 for $247.00
|
|
|||
01-184-595
|
SiliCycle™
SPEP001503BBJ |
60 mg | 3 mL | 200 Pk. |
Pack of 200 for $934.50
|
|
|||
01-184-600
|
SiliCycle™
SPEP001506BB |
60 mg | 6 mL | 30 Pk. |
Pack of 30 for $223.50
|
|
|||
01-184-601
|
SiliCycle™
SPEP001506BBJ |
60 mg | 6 mL | 200 Pk. |
Pack of 200 for $1,135.00
|
|
|||
01-184-858
|
SiliCycle™
SPEP001503C |
100 mg | 3 mL | 50 Pk. |
Pack of 50 for $263.50
|
|
|||
01-184-596
|
SiliCycle™
SPEP001503CJ |
100 mg | 3 mL | 200 Pk. |
Pack of 200 for $994.50
|
|
|||
01-184-597
|
SiliCycle™
SPEP001503CL |
100 mg | 3 mL | 500 Pk. |
Pack of 500 for $2,360.00
|
|
|||
01-184-602
|
SiliCycle™
SPEP001506C |
100 mg | 6 mL | 30 Pk. |
Pack of 30 for $279.50
|
|
|||
01-184-598
|
SiliCycle™
SPEP001503G |
200 mg | 3 mL | 50 Pk. |
Pack of 50 for $310.50
|
|
|||
01-184-821
|
SiliCycle™
SPEP001506G |
200 mg | 6 mL | 30 Pk. |
Pack of 30 for $295.00
|
|
|||
01-184-599
|
SiliCycle™
SPEP001503P |
500 mg | 3 mL | 50 Pk. |
Pack of 50 for $555.00
|
|
|||
01-184-603
|
SiliCycle™
SPEP001506P |
500 mg | 6 mL | 30 Pk. |
Pack of 30 for $398.00
|
|
|||
01-184-806
|
SiliCycle™
SPEP001512P |
500 mg | 12 mL | 20 Pk. |
Pack of 20 for $366.50
|
|
|||
01-184-604
|
SiliCycle™
SPEP001520S |
1 g | 25 mL | 20 Pk. |
Pack of 20 for $660.00
|
|
|||
01-184-796
|
SiliCycle™
SPEP001512U |
2 g | 12 mL | 20 Pk. |
Pack of 20 for $817.50
|
|
|||
01-184-605
|
SiliCycle™
SPEP001520U |
2 g | 25 mL | 20 Pk. |
Pack of 20 for $785.50
|
|
|||
01-184-587
|
SiliCycle™
SPEP0015011A |
10 mg | 1 mL | 100 Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
01-184-591
|
SiliCycle™
SPEP0015031A |
10 mg | 3 mL | 50 Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
- For use with vacuum manifolds and automated SPE equipments
- Tightly packed and homogeneous silica bed for reliability and increased productivity
- Tight particle size distribution and very good packing with no fines, allowing excellent separation
- High recoveries and yields
- Reproducible flow rates lot-to-lot
- Flanged polypropylene tubes
- 20 μm polyethylene frits
Specifications
Polymer | |
Extraction of strongly basic compounds, highly stable in organic solvents | |
85 μm | |
Polystyrene-divinylbenzene WCX | |
Polypropylene (PP) tube, Polyethylene (PE) frits | |
60 Å |
0.85 | |
Vacuum Manifolds or Automated SPE Equipment | |
SiliaPrepX | |
SPE Cartridge | |
Spherical | |
800 m2/g |