Learn More
SiliCycle™ SiliaPrepX™ SPE Cartridges - SCX
SPE Cartridges packed with a polymeric strong cation exchange (SCX) phase, for extraction of weak & medium bases.
$208.50 - $2650.00
Specifications
Adsorbent Type | Polymer |
---|---|
Carbon Load | 8% |
For Use With (Application) | Extraction of weak and medium bases, highly stable in organic solvents |
For Use With (Equipment) | Vacuum Manifolds or Automated SPE Equipment |
Particle Size | 85 μm |
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
01-182-944
|
SiliCycle™
SPEP000501AA |
30 mg | 1 mL | 100 Pk. |
Pack of 100 for $295.00
|
|
|||
01-182-945
|
SiliCycle™
SPEP000501AAL |
30 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,394.00
|
|
|||
01-182-946
|
SiliCycle™
SPEP000501AAM |
30 mg | 1 mL | 1000 Pk. |
Pack of 1000 for $2,650.00
|
|
|||
01-182-951
|
SiliCycle™
SPEP000503AA |
30 mg | 3 mL | 50 Pk. |
Pack of 50 for $208.50
|
|
|||
01-182-952
|
SiliCycle™
SPEP000503AAJ |
30 mg | 3 mL | 200 Pk. |
Pack of 200 for $744.00
|
|
|||
01-182-947
|
SiliCycle™
SPEP000501BB |
60 mg | 1 mL | 100 Pk. |
Pack of 100 for $316.50
|
|
|||
01-182-948
|
SiliCycle™
SPEP000501BBL |
60 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,498.00
|
|
|||
01-182-953
|
SiliCycle™
SPEP000503BB |
60 mg | 3 mL | 50 Pk. |
Pack of 50 for $247.00
|
|
|||
01-182-954
|
SiliCycle™
SPEP000503BBJ |
60 mg | 3 mL | 200 Pk. |
Pack of 200 for $934.50
|
|
|||
01-182-961
|
SiliCycle™
SPEP000506BB |
60 mg | 6 mL | 30 Pk. |
Pack of 30 for $223.50
|
|
|||
01-182-962
|
SiliCycle™
SPEP000506BBJ |
60 mg | 6 mL | 200 Pk. |
Pack of 200 for $1,109.00
|
|
|||
01-182-949
|
SiliCycle™
SPEP000501C |
100 mg | 1 mL | 100 Pk. |
Pack of 100 for $365.50
|
|
|||
01-182-955
|
SiliCycle™
SPEP000503C |
100 mg | 3 mL | 50 Pk. |
Pack of 50 for $263.50
|
|
|||
01-182-956
|
SiliCycle™
SPEP000503CJ |
100 mg | 3 mL | 200 Pk. |
Pack of 200 for $994.50
|
|
|||
01-182-957
|
SiliCycle™
SPEP000503CL |
100 mg | 3 mL | 500 Pk. |
Pack of 500 for $2,360.00
|
|
|||
01-182-963
|
SiliCycle™
SPEP000506C |
100 mg | 6 mL | 30 Pk. |
Pack of 30 for $279.50
|
|
|||
01-182-958
|
SiliCycle™
SPEP000503G |
200 mg | 3 mL | 50 Pk. |
Pack of 50 for $310.50
|
|
|||
01-182-959
|
SiliCycle™
SPEP000503GJ |
200 mg | 3 mL | 200 Pk. |
Pack of 200 for $1,175.00
|
|
|||
01-182-965
|
SiliCycle™
SPEP000506G |
200 mg | 6 mL | 30 Pk. |
Pack of 30 for $295.00
|
|
|||
01-182-966
|
SiliCycle™
SPEP000506GJ |
200 mg | 6 mL | 200 Pk. |
Pack of 200 for $1,465.00
|
|
|||
01-182-960
|
SiliCycle™
SPEP000503P |
500 mg | 3 mL | 50 Pk. |
Pack of 50 for $555.00
|
|
|||
01-182-967
|
SiliCycle™
SPEP000506P |
500 mg | 6 mL | 30 Pk. |
Pack of 30 for $398.00
|
|
|||
01-182-968
|
SiliCycle™
SPEP000512P |
500 mg | 12 mL | 20 Pk. |
Pack of 20 for $344.00
|
|
|||
01-182-969
|
SiliCycle™
SPEP000512S |
1 g | 12 mL | 20 Pk. |
Pack of 20 for $388.00
|
|
|||
01-182-971
|
SiliCycle™
SPEP000520S |
1 g | 25 mL | 20 Pk. |
Pack of 20 for $660.00
|
|
|||
01-182-970
|
SiliCycle™
SPEP000512U |
2 g | 12 mL | 20 Pk. |
Pack of 20 for $817.50
|
|
|||
01-182-972
|
SiliCycle™
SPEP000520U |
2 g | 25 mL | 20 Pk. |
Pack of 20 for $785.50
|
|
|||
01-182-943
|
SiliCycle™
SPEP0005011A |
10 mg | 1 mL | 100 Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
01-182-950
|
SiliCycle™
SPEP0005031A |
10 mg | 3 mL | 50 Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
- For use with vacuum manifolds and automated SPE equipments
- Tightly packed and homogeneous silica bed for reliability and increased productivity
- Tight particle size distribution and very good packing with no fines, allowing excellent separation
- High recoveries and yields
- Reproducible flow rates lot-to-lot
- Flanged polypropylene tubes
- 20 μm polyethylene frits
Specifications
Polymer | |
Extraction of weak and medium bases, highly stable in organic solvents | |
85 μm | |
Polystyrene-divinylbenzene SCX | |
Polypropylene (PP) tube, Polyethylene (PE) frits | |
60 Å |
8% | |
Vacuum Manifolds or Automated SPE Equipment | |
SiliaPrepX | |
SPE Cartridge | |
Spherical | |
800 m2/g |