Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Thiol
SPE Cartridges packed with SiliaMetS Thiol, to lower residual metal concentration of various metal complexes (Ag, Hg, Os, Pd & Ru).
Supplier:  SiliCycle™ SPER51030B06S
 View more versions of this product
Catalog No. 01-184-844
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Lower residual metal concentration in Ag, Hg, Os, Pd, Ru
40 to 63 μm
Thiol
6 mL
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
12.7 x 77.8 mm
12.7 mm
1 g
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep
SPE Cartridge
Yes
Irregular
50 Pk.
77.8 mm
Product Suggestions
Product Suggestions