Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gels, Cyanoborohydride (CBH), 40 to 63 μm, 60 Å
Our Cyanoborohydride (Si-CBH) is very useful in reductive amination and in the reduction of imines and aldehydes.
$87.80 - $4000.00
Specifications
For Use With (Application) Reductive Amination and Reduction of Imines and Aldehydes
Particle Size 40 to 63 μm
Product Type Bulk Functionalized Irregular Silica Gel
Particle Shape Irregular
Pore Size 60 Å
View More Specs
Products 8
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-184-430
View Documents
SiliCycle™
R66730B10G
10 g
Each for $87.80
Add to Cart
01-184-388
View Documents
SiliCycle™
R66730B25G
25 g
Each for $168.30
Add to Cart
01-184-389
View Documents
SiliCycle™
R66730B50G
50 g
Each for $314.50
Add to Cart
01-184-420
View Documents
SiliCycle™
R66730B100G
100 g
Each for $515.50
Add to Cart
01-184-390
View Documents
SiliCycle™
R66730B250G
250 g
Each for $1,171.00
Add to Cart
01-184-391
View Documents
SiliCycle™
R66730B500G
500 g
Each for $2,220.00
Add to Cart
01-184-392
View Documents
SiliCycle™
R66730B1KG
1 kg
Each for $4,000.00
Add to Cart
01-184-387
View Documents
SiliCycle™
R66730B5G
5 g Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
SiliaBond™ Cyanoborohydride (Si-CBH) is the silica-bound equivalent of sodium cyanoborohydride. Bound cyanoborohydride is very useful in reductive amination and in the reduction of imines and aldehydes. Cyanide contamination of products is a concern when using the solution phase equivalent. This problem is minimized with the use of silica-bound materials, since the toxic cyanide residue remains on the silica. Additionally, it may be used to reduce α, β-unsaturated carbonyl to the corresponding unsaturated alcohol.
To determine if any cyanide ion was leaching from the silica, 1 g of SiliaBond™ Cyanoborohydride was washed in 10 mL of methanol for 24 hours. Cyanide strips indicated less than 3 ppm in each test performed.
Specifications
Specifications
Reductive Amination and Reduction of Imines and Aldehydes
Bulk Functionalized Irregular Silica Gel
60 Å
Cyanoborohydride (CBH)
40 to 63 μm
Irregular
SiliaBond™