Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Public Seating™ Signature Science Lab Table, Textured Black Legs
SCI_ED

NPS Signature Frame Adjustable Height Science Lab Tables have perfect chemistry with any science classroom.

Supplier:  National Public Seating™ SLT42460H

 View more versions of this product

Catalog No. S83683


Add to Cart

Description

Description

  • Round Legs have (2) 3/8 in. Diameter in Textured Black
  • Legs Adjust Smoothly and Easily in 3/4 in. Increments – from 27 to 42 in. Height
  • Adapts for ADA Requirements
  • Easy Assembly, With Pre-Drilled Pilot Holes
  • Patented Mechanism Allows Table to Rise Effortlessly and Automatically Lock At Your Height Setting Requirement
  • Weight Capacity: 500 lb.
  • Oversized Matching Floor Glides are Standard
  • 10 Year Warranty
Specifications

Specifications

Lab Table
MAS2
Science Classrooms
500 lb.
Black
60 x 24 x 27 to 42 in.
HPL
10 years
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit