Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Diagnostics™ Formalin Solution, Neutral Buffered, 10%
General purpose histological fixative
Supplier: Sigma Diagnostics™ HT5011284L
Description
- Physical form: Neutral buffered solution
Specifications
Colorless | |
4L | |
General purpose histological fixative | |
Solution |
Formalin | |
Formaldehyde 37 to 40% 100mL/L, distilled or deionized Water 900mL/L, Sodium Phosphate monobasic 4g/L, Sodium Phosphate, dibasic (anhydrous) 6.5g/L | |
6.90 - 7.10 |
Safety and Handling
ShelfLife : Expiry date on the label
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction