Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Diagnostics™ Eosin Y Solution, aqueous
General purpose cytoplasmic counterstain
$79.60
Specifications
Type | Aqueous Eosin Y Stain |
---|---|
Format | Solution |
For Use With (Application) | Used with hematoxylin and eosin staining |
Product Type | Eosin Y solution, aqueous |
Storage Requirements | 18–26°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
SDHT110216
|
Sigma Diagnostics™
HT110216500ML |
500mL |
Each of 1 for $79.60
|
|
|||||
Description
- Certified Eosin Y, 0.5% (w/v) in water. Not acidified.
Specifications
Aqueous Eosin Y Stain | |
Used with hematoxylin and eosin staining | |
18–26°C |
Solution | |
Eosin Y solution, aqueous |
Safety and Handling
ShelfLife : Expiry date on the label.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title