Learn More
Perkin Elmer US LLC SiC Abrasive Pad - Qty. 100
Supplier: Perkin Elmer US LLC L1271021
PerkinElmer's Diffuse Reflectance Accessory (L1200351/L1600111/L1280323) comes with abrasive sampling tools that allow you to collect ready-to-analyze dust samples from hard-surfaced solid materials. The Diffuse Reflectance Multi-Purpose Sample Holder (L1201865), designed to hold either an abrasive stick or pad, easily slides into the diffuse reflectance accessory. The holder can accommodate up to two sticks or two pads; the first tool with the sample dust goes in its sample position, the second tool, in its reference position.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.