Learn More
Xylem Inc SI VZ 7010 PTFE TUBING SET
Supplier:  Xylem Inc 285420158
Encompass_Preferred
SI VZ 7010 PTFE tubing set Diai 1,5 mm, suitable forthermostate type: CT 72/2 and CT 72/4. Forviscometers of type series: 501.., 525.., 526.., 530..,532.., 536.., 537.., 538.., 552.. and 554
Catalog No. NC2595670