Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOTRAX™ Shoe Sanitizer System
These shoe sanitizing systems provide an effective solution to minimize the spread of contaminants tracked on the bottoms of shoes.
$511.50 - $685.50
Catalog Number | Mfr. No. | Description | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Dimensions (L x W) | Price | Quantity & Availability | ||||
19-111-132
|
NOTRAX™
355S002500 |
Shoe Disinfecting and Drying System (2-Zone) | 52.5 x 25.5 in. |
Each for $511.50
|
|
||||
19-111-135
|
NOTRAX™
355S002700 |
Shoe Cleaning, Disinfecting, and Drying System (3-Zone) | 108.5 x 27 in. |
Each for $685.50
|
|
||||
Description
- Simple, safe and effective shoe disinfecting system to limit the spread of contaminants on floors
- Labels clearly instruct how to sanitize shoes
- Ideal for entrances to schools, businesses, facilities, and more
- Low profile bevels for easy walk on/off to minimize trip hazards
- Designed as a zoned system with completely contained and secured components
- Carpet mat inserts are disposable and replaceable for quick and easy cleaning
- Available in 2-zone (disinfecting, drying) and 3-zone (cleaning, disinfecting, drying) options
- Each system cleans and sanitizes shoe soles, then quickly dries them the moment you step out
- Mat inserts are adhesive backed and replaceable for easy cleaning
- Carpet mats are made from recycled plastic bottles
- System is lightweight and easy to set up and move
- Vinyl backed to prevent slipping
Specifications
Black | |
Recycled Plastic | |
Entrance Mat |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title