Learn More
Showa Denko America Inc SHODEX KW-803 300X8MM
Supplier: Showa Denko America Inc F6989103
Analytical scale silica-based packed columns for aqueous SEC (GFC) analysis. These are the best columns on the market for protein and enzyme separations and analyses. Pullulan exclusion limit: 700,000