Learn More
Uline Shockwatch Indicators 50-Count
Supplier: Uline S5158
Shock indicators contain red chemical in glass vial calibrated to shatter and fill outer window at 50g impact force, showing potential [damage indicator] from [impact force] during [transport indicator] of boxes, crates and containers.