Learn More
Bioworld Sheep serum, sterile filtered, 0.2 absolute, heat inactivated, 100 mL
Supplier:  Bioworld 306111681
Encompass_Preferred
All animals are certified of U.S. origin.An exceptional quality serum.Physiochemical constituents are measured to ensure lot-to-lot consistency.Sterility tested.Endotoxin testing upon request.Mycoplasma testing upon request.Sterile filtered to 0.2 µm.Storage Conditions: -20°C + 5°C.Requires Dry Ice ShippingUsage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
Catalog No. 50-254-241