Learn More
Colorado Serum Co SHEEP RBC IN ALSEVERS 60ML
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Colorado Serum Co 31112
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
SHEEP RBC IN ALSEVERS, 60ml; 50% mix of whole blood and alsevers solution. Alsevers solution (which is made in our laboratory and sterility tested) acts as an anti-coagulant and preservative and contains sodium citrate and glucose.