Learn More
Colorado Serum Co Sheep Alsevers, 100 mL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Colorado Serum Co 31113
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Alsevers solution, which is made in our laboratory and tested for sterility, acts as an anticoagulant and preservative. It contains sodium citrate and glucose.