Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sterilis Solutions Sharps Container, 4 Gallon Cylinder
Sterilis Solutions Sharps Container, 4 Gallon Cylinder is a puncture-resistant sharps container with wide circular opening for easy loading of needles.
$288.50 - $963.00
Specifications
Material | Polyethylene |
---|---|
Product Type | Sharps Container |
Capacity (Metric) | 15 L |
Capacity (English) | 4 gal. |
Color | Translucent Red |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-777-026
|
Sterilis Solutions
3823310001 |
10 Pk. |
Pack of 10 for $288.50
|
|
|||||
23-777-027
|
Sterilis Solutions
3823310002 |
20 Pk. |
Pack of 20 for $535.50
|
|
|||||
23-777-025
|
Sterilis Solutions
38233100 |
40 Pk. |
Pack of 40 for $963.00
|
|
|||||
Description
- General-purpose sharps container with clear locking lid and marked with biohazard label
- Compliant with FDA and OSHA standards for sharps and biohazard waste collection
- Puncture-resistant with wide circular opening for vertical or horizontal loading of needles
- Made from transparent red puncture-resistant polypropylene that can be autoclaved or incinerated
- Rotating handle for easy transport and handling
- Nests to conserve storage space
Specifications
Polyethylene | |
15 L | |
Translucent Red | |
Yes | |
Yes | |
Yes | |
Yes | |
13 in. |
Sharps Container | |
4 gal. | |
11.37 x 13 in. | |
330.1 mm | |
Cylindrical | |
Sterile | |
Autoclavable |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title