Learn More
Cole-Parmer Sharpie SAN-13601 Industrial marker with fine tip, black
Supplier: Cole-Parmer 0996900
Super-permanent black ink formulated for laboratory, industrial, and commercial use. Ink resists most chemical washes, steam, and heat up to 260°C (500°F).