Learn More
Andwin Scientific Tissue-Tek Accu-Edge Blades Sharp/Blunt 5/case
Supplier: Andwin Scientific 4796
The Tissue-Tek Accu-Edge replaceable Sharp/Blunt blade is an ideal tool for performing anatomical sectioning of tissues. The blades can be easily inserted and removed, which removes the time, expense, and incovenience of sharpening. This product is autoclavable and is sold in a case of 5.
This item is not returnable.
View return policy