Learn More
Epredia™ Shandon™ Rapid-Chrome Papanicolaou Staining Kit
Achieve rapid results of fine needle aspirations or high-risk specimens with the Epredia™ Shandon™ Rapid-Chrome Papanicolaou Staining Kit.
Supplier: Epredia™ 9990020
Description
Includes: five-slide holder, portable tray, rinse water, Gill 2 Hematoxylin, bluing reagent, EA-50, 100% alcohol, OG-6, 95% alcohol, Xylene, mounting mediumSpecifications
Papanicolaou Stain | |
Papanicolaou staining kit; Five-slide holder |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov