Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Shandon™ Grosslab™ Filter Sets
Each filter set contains a formaldehyde neutralizing (Potassium Permanganate) filter along with a fine particle (Charcoal) filter suitable for Grosslab Jr. and Grosslab Sr. workstations.
Supplier: Epredia™ 9990850
Description
- Contains a Formaldehyde Neutralizing (Potassium Permanganate) Filter along with a Fine Particle (Charcoal) Filter
- Filter Size: 42.7 x 28.6 x 5.1 cm
- For Use With Grosslab Senior Series 8700 and 9700
Specifications
Filter Set | |
Contains a Formaldehyde Neutralizing (Potassium Permanganate) Filter along with a Fine Particle (Charcoal) Filter | |
Grosslab Senior Series 8700 and 9700 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction