Learn More
Epredia™ Shandon™ Dual Blade Manual Bone Saw, Replacement Blades, Fine
The Epredia™ Shandon Dual Blade Manual Bone Saw makes simultaneous parallel cuts to remove a slice of bone for processing.
Supplier: Epredia™ 1001799
Description
Operator places the bone between the movable upper jaws and the fixed lower jaw of the bone saw to hold the specimen securely.- Cuts a wide range of bones easily, including difficult femoral heads
- Manual design inhibits aerosols as compared with electric saws
- Easy to clean, high-quality hacksaw with dual blades produces consistent cuts
- 7 × 9in.W (17.8L × 22.9cm) unit accommodates specimens up to 7 × 9in.W (7.6L × 10.2cm) in cross section and of indefinite length
Includes:One set each of coarse and fine blades; replacement blade sets include links.
Specifications
Dual Blade Manual Bone Saw | |
Replacement blade set include links | |
17.8 cm | |
22.9 cm |
Replacement Blades, Fine | |
7 in. | |
9 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.