Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Shandon™ Dissecting Tray for Raised Dissecting Boards, 12x 11 in.
Epredia™ Shandon Dissecting Tray for Raised Dissecting Boards feature ample space for drainage and instruments.
Supplier: Epredia™ 35614
Description
- Four embossments on tray bottom hold Raised Dissecting Boards 356 and 35614
Specifications
Dissecting Tray | |
Small | |
11 in. |
Polyethylene | |
12 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction