Learn More
Epredia™ Shandon™ ColorFrost™ Plus Slides
Epredia™ Shandon ColorFrost Plus Slides are positively charged for improved cellular adhesion.
Supplier:  Epredia™ 9991011
 View more versions of this product
Catalog No. 99-910-11
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Electrostatically attract tissue sections and cytology preparations, binding them to the slide
  • Eliminate the need for special adhesives or protein coatings
  • No background staining
  • Available in a variety of colors
Specifications
Specifications
Microscope Slide
Glass
100/Cs.
Yes
Burnt Orange