Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Shandon™ Bone Chisel 0.5″ beveled edge, One Bevel, 0.5 in. (1.3cm) edge, standard, 6.5 in. (16.5cm)

Make precision cuts and shavings with the Epredia™ Shandon™ Bone Chisel, guaranteed to be free from defects in material and worksmanship.

Supplier:  Epredia™ 6002

Catalog No. 60-02


Add to Cart

Description

Description

Features:

  • Length: 6.5 in. (16.5cm)
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
Specifications

Specifications

Bone Chisel
Shandon™
0.5 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit