Learn More
Cole-Parmer Stuart Wrist-Action Flask Shaker, 230 V
Supplier: Cole-Parmer 5190002
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The variable-speed shaker features two side arms with adjustable clamps capable of holding eight flasks or bottles up to 500 mL capacity. Use for prolonged continuous shaking or set the built-in analog timer from 10 to 60 minutes and unit automatically turns off when done. Electronic feedback controller ensures a constant speed, irrespective of load.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.