Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva SFX-Insect™ Cell Culture Media
Designed to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. HyClone™ SFX Insect™ Cell Culture Media is a protein-free media that support cell culture growth.
Supplier: Cytiva SH30350.02
This item is not returnable.
View return policy
Description
- Protein free media
Specifications
cGMP | |
Powder | |
36 Months | |
Non-sterile | |
No Sodium Bicarbonate, No Protein, No Phenol Red, No HEPES |
2° to 8°C, away from light and moisture | |
Insect Cell Medium | |
Non-sterile | |
Glutamine, Poloxamer 188 | |
5 L |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title