Learn More
Expression Systems Sf21 cells, Frozen in ESF 921, 50M cells/vial
Supplier: Expression Systems 94003F
Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
The Sf21 insect cell line (IPLB-Sf-21-AE) is derived from ovarian cells of Spodoptera frugiperda. Sf21 cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture.
• Recombinant protein expression from a variety of expression platforms
• Good growth in adherent or suspension culture
• Quality and performance testing
To download Instructions for Use and/or COA please visit: www.expressionsystems.com
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.