Learn More
Raven Environmental Products Raven Settlometer Kit, 1L Volume, Acrylic
Supplier: Raven Environmental Products C10102
The Raven Settlometer Kit is used to evaluate and troubleshoot activated sludge settling characteristics. Use with mixed liquor samples to determine the behavior of the activated sludge in the secondary clarifier. Use with aerobic digester samples to evaluate supernatants and separation periods for sludge wasting. Although narrower than other Settleometers, settling is not distorted and is easy to carry or transport. Marked in % and Settled Sludge Volume (SSV). 1 liter, 4 in. diameter, 8 in. tall. Comes with snap on lid and wide mouth paddle.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.