Learn More
Cole-Parmer Boekel Stopper Hole Boring Set, Brass; 9 Borers/Set
Supplier: Cole-Parmer 0629891
Convert your solid stopper into holed-stopper with these easy-to-use borer sets. All borers come with a steel punch to make boring easier. Borer sharpener (sold separately as part number 06298-94) keeps your borer in top condition and sharpens borers up to 25mm in diameter.