Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Set of 4 dNTPs (100mm)
Offers maximum flexibility in preparation of reaction mixes. MP Biomedicals Set of 4 dNTPs (100mm) includes four separate vials of highly purified sodium salts of dATP, dCTP, dGTP and dTTP. These solutions achieve sensitive and consistent PCR results.
$433.00 - $1252.00
Specifications
Purity or Quality Grade | 99+% |
---|---|
For Use With (Application) | Standard PCR, Real Time PCR (qPCR), RT-PCR |
Concentration | 100 mM |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP1NTACG100
|
MP Biomedicals™
11NTACG100 |
4 x 25 μmol |
Each for $433.00
|
|
|||||
MP1NTACG111
|
MP Biomedicals™
11NTACG111 |
4 x 100 μmol |
Each for $1,252.00
|
|
|||||
Description
This product allows the addition of each dNTP to be altered as required for specific experiments, such as:
- When adding a radioactive nucleotide for labeling by PCR
- To increase the chance of non-specific mutations
- In PCR, real-time PCR, cDNA synthesis and DNA sequencing
Specifications
99+% | |
Standard PCR, Real Time PCR (qPCR), RT-PCR | |
100 mM |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title