Learn More
Crescent Chemical Co Inc SERVALYT™ 4-7
Supplier: Crescent Chemical Co Inc SE42948.02
SERVALYT™ 4-7 40 % (w/v) solution in water SERVALYT™ Carrier Ampholytes (40 % w/v in water). Standard quality (analytical grade) for general use in IEF. Size - 25ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42948.02