Learn More
Crescent Chemical Co Inc SERVALYT™ 3-10
Supplier: Crescent Chemical Co Inc SE42940.01
SERVALYT™ 3-10 40 % (w/v) solution in water SERVALYT™ Carrier Ampholytes (40 % w/v in water). Standard quality (analytical grade) for general use in IEF. Size - 10ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42940.01