Learn More
Crescent Chemical Co Inc SERVA TRIS-GLYCINE NATIVE BUFF
Supplier: Crescent Chemical Co Inc SE42530.01
SERVA Tris-Glycine native electrophoresis buffer (10x) Running buffer for native PAGE. Supplied as 10 x concentrate. Contains 250 mM Tris, 1.92 M glycine. Size - 1L Storage Conditions - +2 °C TO +8 °C Catalog Number - 42530.01