Learn More
Crescent Chemical Co Inc SERVA PROTEINSTAIN FLUO-R POWD
Supplier: Crescent Chemical Co Inc SE35090.01
SERVA ProteinStain Fluo-R, powder SERVA ProteinStain Fluo-R is a highly sensitive fluorescent dye for detection of proteins in e. g. SDS PAGE, native PAGE or 2D gels. The dye does not interfere with immunodetection. Therefore you can stain your gel with SERVA ProteinStain Fluo-R