Learn More
Gemini Bio Products SERUM NEWBORN CALF 500ML
Supplier: Gemini Bio Products 100504/500
Newborn calf serum will support a wide variety of established cell lines and primary cultures which might otherwise require fetal bovine serum. Empirical testing is the most reliable method for determining usefulness for a particular line.