Learn More
Cytiva SERIES S SENSOR CHIP CM7 3/PK
Supplier: Cytiva 29147020
Series S Sensor Chip CM7, pack of 3, for use when achieving the required immobilization level is a challenge. Suitable for small molecule work and fragment-based screening.