Learn More
Cytiva SERIES S SENSOR CHIP CM7
Supplier: Cytiva 28953828
Use when achieving the required immobilization level is a challenge. Suitable for small molecule work and fragment-based screening.