Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BINDER™ Series FED Avantgarde.Line Drying and Heating Chambers PROMO
BINDER™ Series FED Avantgarde.Line Drying and Heating Chambers feature advanced timing functions, controllable fan, and easy to control temperature and convection conditions.
Supplier: BINDER™ 90100294PM
Description
- Up to 30% lower energy consumption compared to conventional units on the market
- High temperature accuracy thanks to APT.line™ technology
- Adjustable fan speed
- controller with LCD display and enhanced time functions
- Electromechanical control of the exhaust air flap
- 2 chrome-plated racks
- Units up to 115 liters are stackable
- Class 2 integrated independent adjustable temperature safety device (DIN 12880) with visual alarm
- Ergonomic handle design
- Ethernet interface
- USB port for recording data
Specifications
116 L | |
Drying and Heating Chamber | |
30 kg | |
2 std. / 5 max. | |
605 x 710 x 735 mm |
Ambient +10° to 300°C | |
1 door | |
FED115UL-120V | |
385 x 550 x 550 mm | |
120 V, 60 Hz, 12.5 A |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction