Learn More
BINDER™ Series E Classic.Line - Drying and Heating Chamber with Mechanical Adjustment
BINDER™ Series E Classic.Line Drying and Heating Chamber with Mechanical Adjustment is preferred for tasks in basic research, human medicine, and veterinary medicine.
Supplier: BINDER™ 90100106
Description
- Adjustable exhaust air flap
- Hydro-mechanical thermostat
- Timer 0 to 120 minutes
- 2 chrome-plated racks
Specifications
1 cu. ft. | |
28 L | |
1 door | |
E028-120V | |
400 x 250 x 280 mm | |
48.5 lb. | |
120 V, 60 Hz | |
0.8 kW | |
60 Hz |
E028-120V, Series E Classic.Line - Drying and Heating Chamber with Mechanical Adjustment, 28 L, 120 V 1∼ 60 Hz | |
60°C to 230°C | |
10 kg | |
2 std. / 4 max. | |
425 x 580 x 405 mm | |
22 kg | |
120 V | |
3.5 A |