Learn More
Seracare Life Sciences Inc SERACON II 1 LITER
Supplier: Seracare Life Sciences Inc 18000027
Material derived from human plasma collected in FDA-licensed facilities and tested in accordance with FDA guidelines for viral markers. The plasma is tested and found non-reactive for HBsAg, anti-HIV1/2, anti-HCV, anti-HBc, anti-HTLV I/II, HIV-1 RNA, HCV RNA, and HBV DNA. Donors are also routinely screened for syphilis negative status prior to donating. The plasma is processed in an FDA-registered, ISO-certified 9001 and 13485 facility and retains lot-to-lot consistency with total protein (>6.0 g/dL), fibrinogen (<25 mg/dL), and cholesterol/triglycerides (<20 mg/dL). Processing includes: • Pooling into batches up to 1,000 Liters • Removal of fibrinogen • Removal of lipids • Charcoal stripped for hormone depletion • 0.2 μm filtered and dialyzed to mimic a serum-like state
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.