Learn More
Chemglass Life Sciences Septum Holder, M27 Thread, for Bio Reactors
Supplier: Chemglass Life Sciences CLS138509
Septum Holder, M27 Thread, for Bio Reactors. The septum holder is supplied with 1 silicone rubber septa and can be used as a universal addition port for fluids or gas. Replacement septa are available.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.