Learn More
Chemglass Life Sciences 8-425 Replacement Septa, Minum-Ware
SDP
Supplier:  Chemglass Life Sciences MW10401
Encompass_Preferred
8-425 Replacement Septa, Minum-Ware. Replacement septa for use with MW-102 open-top screw caps on Minum-Ware glassware components. Septa are made of silicone rubber with one side having a RED PTFE face, while the opposite side has an ultrathin covering of polypropylene to prevent needle coring when penetrating septa. Sold in packages of 12 or 144 each.
Catalog No. 50-872-959