Learn More
Hach Company Analog Flow Sensor Input Module for sc200 Universal Controller
Supplier: Hach Company 9012700
The Flow Sensor module for the SC200 is a user installed module that allows connection of flow meters to the controller. This module will work with analog flow sensors and is specifically built to be used with the flow paddle wheel sensor and the ultrasonic flow sensor. See manual for additional connection information.