Learn More
Cytiva Sensor Chip Protein G, Pack of 1
Supplier: Cytiva 29179316
A Biacore Extend product used for oriented capture or binding of a wide range of mammalian and all human antibody subclasses.