Learn More
Electron Microscopy Sciences Sensidyne Gilian BDXII Pump
Supplier: Electron Microscopy Sciences 6524112
The Gilian BDX II Abatement Air Sampler from Sensidyne is a dependable, low cost personal sampling pump. It is a "no-frills" model designed for use with asbestos and lead sampling. Meets NIOSH 7400, 7082, 7105 and 7300 specifications. Flow rates 500-3000 cc/min (0.5-3 LPM). Comes with rechargeable battery pack system and built-in rotameter and flow adjust. It is compact, lightweight, and quiet. Full one year warranty.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.