Learn More
Sensidyne™ Colorimetric Gas Detector Tubes: Isopentyl Alcohol
Supplier: Sensidyne™ 209U
Specifications
Isopentyl Alcohol | |
10 Tubes/Box | |
0°C to 40°C |
5 to 100ppm | |
32°F to 104°F | |
Standard tube |