Learn More
Sensidyne™ Colorimetric Gas Detector Tubes: Ethyl Ether
Supplier: Sensidyne™ 107U
Specifications
Ethyl Ether | |
10 Tubes/Box | |
10° to 40°C |
20 to 400ppm | |
50°F to 104°F | |
Standard tube |