Learn More
Perkin Elmer US LLC Semi-Demountable Cell Window - BaF2. 4 mm, Qty. 2
Supplier: Perkin Elmer US LLC L1271194
Semi-Demountable Cell Windows are rectangular windows and come in a variety of window materials. Each window measures 41 (H) x 23 mm (W), and they come in pairs. These windows require, but do not include, the Universal Demountable Cell (L1270986).