Learn More
World Precision Instrument Semi Micro Concentric Bipolar Electrodes, Rhodes Style Tip-SSM321RC100
Supplier: World Precision Instrument SSM321RC100
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.